Product Details
- SNP ID
-
rs13475882
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:58903699 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGCTTTTTCCGTTCTCCTGGGGATC[A/G]GGGGTCTTGACCACCCTGGCTATTC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Trak2
PubMed Links
Gene Details
- Gene
- Trak2
- Gene Name
- trafficking protein, kinesin binding 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_172406.3 |
2991 |
UTR 3 |
|
|
NP_765994.2 |
View Full Product Details