Product Details

SNP ID
rs6237258
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.5:129965381 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TAAGACAAAGGCTTTAGATGTGAAC[C/T]GAAGGCATCACCATTTGACTTGCAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Vkorc1l1 PubMed Links

Gene Details

Gene
Vkorc1l1
Gene Name
vitamin K epoxide reductase complex, subunit 1-like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001327.2 Intron NP_001001327.1
NM_001286382.1 Intron NP_001273311.1
NM_027121.4 Intron NP_081397.1

View Full Product Details