Product Details
- SNP ID
-
rs13483068
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:64025872 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGAGCCAGTCTGTGTTCTTAGACT[C/T]AACAGAGTATCCAGTGCTGTGTCAT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Fer
PubMed Links
Gene Details
- Gene
- Fer
- Gene Name
- fer (fms/fps related) protein kinase
View Full Product Details