Product Details

SNP ID
rs13475927
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.1:74902192 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
ACACCAGCAAACAGTCAGGATGCTG[C/T]ATCCCAGAGCAGAGCACGTCCCCTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Ccdc108 PubMed Links

Gene Details

Gene
Ccdc108
Gene Name
coiled-coil domain containing 108
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039495.1 5606 Missense Mutation NP_001034584.1
Gene
Cryba2
Gene Name
crystallin, beta A2
There are no transcripts associated with this gene.

Gene
Mir375
Gene Name
microRNA 375
There are no transcripts associated with this gene.

View Full Product Details