Product Details

SNP ID
rs13482623
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.15:66920653 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TGAAGACAGAAGTCGACCAGCCATT[C/T]AGAAATGGGGATCTATTTAGTAGTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Ndrg1 PubMed Links

Gene Details

Gene
Ndrg1
Gene Name
N-myc downstream regulated gene 1
There are no transcripts associated with this gene.

Gene
Wisp1
Gene Name
WNT1 inducible signaling pathway protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018865.2 2476 UTR 3 NP_061353.1

View Full Product Details