Product Details

SNP ID
rs13478155
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.5:27403159 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TGTCCTCTGGTGTAGCCCCCTCCCT[C/T]GGAGGTAGTTCCCTGTACCTCAGGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Dpp6 PubMed Links

Gene Details

Gene
Dpp6
Gene Name
dipeptidylpeptidase 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136060.2 Intron NP_001129532.1
NM_001198886.1 Intron NP_001185815.1
NM_010075.2 Intron NP_034205.1
NM_207282.3 Intron NP_997165.2

View Full Product Details