Product Details

SNP ID
rs13483123
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.17:80371400 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TCCACTCAGAAGAAATGCTCCAACC[C/T]TATCCTTCCTCTTCCAGTTCCGCCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Arhgef33 PubMed Links

Gene Details

Gene
Arhgef33
Gene Name
Rho guanine nucleotide exchange factor (GEF) 33
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145452.1 1719 Silent Mutation NP_001138924.1
Gene
Gm10190
Gene Name
predicted gene 10190
There are no transcripts associated with this gene.

View Full Product Details