Product Details
- SNP ID
-
rs6193233
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:64845999 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCACGTCAGGTTAAGGGCAGGGACA[C/G]CAAGTTTAGAGCTTTGATTTAATTT
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
Hmbox1
PubMed Links
Gene Details
- Gene
- Hmbox1
- Gene Name
- homeobox containing 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_177338.5 |
|
Intron |
|
|
NP_796312.2 |
View Full Product Details