Product Details

SNP ID
rs13483588
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.19:29628935 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GAAAACAACGGACCCGGGAAGTACA[C/T]GAGCAGACACCTCTTCCCACCCCAC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Ermp1 PubMed Links

Gene Details

Gene
Ermp1
Gene Name
endoplasmic reticulum metallopeptidase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001081213.1 Intron NP_001074682.1

View Full Product Details