Product Details
- SNP ID
-
rs13476578
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:74704654 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGTCTCTGCTGGGCAGATCCAGGAG[C/T]CTTGCTGGAGGAGAGACCGTCTGCT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Hoxd10
PubMed Links
Gene Details
- Gene
- Hoxd10
- Gene Name
- homeobox D10
There are no transcripts associated with this gene.
- Gene
- Hoxd3
- Gene Name
- homeobox D3
There are no transcripts associated with this gene.
- Gene
- Hoxd3os1
- Gene Name
- homeobox D3, opposite strand 1
There are no transcripts associated with this gene.
- Gene
- Hoxd8
- Gene Name
- homeobox D8
There are no transcripts associated with this gene.
- Gene
- Hoxd9
- Gene Name
- homeobox D9
There are no transcripts associated with this gene.
View Full Product Details