Product Details
- SNP ID
-
rs13478702
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:33839014 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGAAGAGAAACTTGTCATCAGTGC[A/T]TCTTGGGCAAAGGATGATGATATCA
- Phenotype
-
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
Exoc4
PubMed Links
Gene Details
- Gene
- Exoc4
- Gene Name
- exocyst complex component 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_009148.3 |
1917 |
Silent Mutation |
|
|
NP_033174.2 |
View Full Product Details