Product Details

SNP ID
rs13481907
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.13:81670469 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TCACACGGACACTGTAATTCGTCAA[G/T]ATCTATTCCTTAAAAAAAGGAATCC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
Lysmd3 PubMed Links

Gene Details

Gene
Lysmd3
Gene Name
LysM, putative peptidoglycan-binding, domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030257.1 1705 UTR 3 NP_084533.1
Gene
Polr3g
Gene Name
polymerase (RNA) III (DNA directed) polypeptide G
There are no transcripts associated with this gene.

View Full Product Details