Product Details

SNP ID
rs6239033
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.11:96955941 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TCCCGCAGTCCGGGATGTGGCACAC[A/G]TGCTTCTTCTTGCCCTGCTCCCCAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
D030028A08Rik PubMed Links

Gene Details

Gene
D030028A08Rik
Gene Name
RIKEN cDNA D030028A08 gene
There are no transcripts associated with this gene.

Gene
Sp2
Gene Name
Sp2 transcription factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080964.1 1633 Silent Mutation NP_001074433.1
NM_030220.3 1633 Silent Mutation NP_084496.2

View Full Product Details