Product Details

SNP ID
rs13480820
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.10:127174816 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
ATTGCCATTTCTCACAAAGTCCACA[C/T]GGCTGATGTGCCAGAGCTGCGGAAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Arhgef25 PubMed Links

Gene Details

Gene
Arhgef25
Gene Name
Rho guanine nucleotide exchange factor (GEF) 25
There are no transcripts associated with this gene.

Gene
B4galnt1
Gene Name
beta-1,4-N-acetyl-galactosaminyl transferase 1
There are no transcripts associated with this gene.

Gene
Slc26a10
Gene Name
solute carrier family 26, member 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_177615.3 1511 Missense Mutation NP_808283.2

View Full Product Details