Product Details

SNP ID
rs13478132
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.5:21494675 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
AATTAAAGGGAAGACTGGGGAATGA[C/T]GTGAAAAAAAAAGGAGTTCTCTTTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Fam185a PubMed Links

Gene Details

Gene
Fam185a
Gene Name
family with sequence similarity 185, member A
There are no transcripts associated with this gene.

Gene
Fbxl13
Gene Name
F-box and leucine-rich repeat protein 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199632.1 Intron NP_001186561.1
NM_177076.4 Intron NP_796050.2

View Full Product Details