Product Details
- SNP ID
-
rs13477725
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:57613710 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAAATGAATTTAGAGTGCAGCTCTG[C/T]TTGTGGTTTTTCAGCCGTAAGTATT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Palm2
PubMed Links
Gene Details
- Gene
- Palm2
- Gene Name
- paralemmin 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_172868.3 |
|
Intron |
|
|
NP_766456.1 |
View Full Product Details