Product Details
- SNP ID
-
rs3679593
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:27120050 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGATTTCCTAAGGTTTCATTTAGTT[G/T]TACAGCTTTTTCATTTATGGCTATG
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
Lama2
PubMed Links
Gene Details
- Gene
- Lama2
- Gene Name
- laminin, alpha 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_008481.2 |
5226 |
Missense Mutation |
|
|
NP_032507.2 |
View Full Product Details