Product Details
- SNP ID
-
rs6352447
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:26551479 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTATTCGGGATGGGTCCCGTCTGGC[A/G]CCAAAGTTTGACTCTGGGCTTTAAT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Fbxl7
PubMed Links
Gene Details
- Gene
- Fbxl7
- Gene Name
- F-box and leucine-rich repeat protein 7
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_176959.3 |
|
Intron |
|
|
NP_795933.2 |
View Full Product Details