Product Details
- SNP ID
-
rs13480470
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:4637805 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAATCGCGACTCACCATGTGATTAT[T/C]ATGCATTGCCATGGGATGCTGCTAA
- Phenotype
-
- Polymorphism
- T/C, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Esr1
PubMed Links
Gene Details
- Gene
- Esr1
- Gene Name
- estrogen receptor 1 (alpha)
There are no transcripts associated with this gene.
View Full Product Details