Product Details
- SNP ID
-
rs6385006
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:32529785 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATCCAGATGTTCTCTCTTCCTTGCG[A/G]AGCCAATGCCACTGGGTGGCCACTG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Gypc
PubMed Links
Gene Details
- Gene
- Gypc
- Gene Name
- glycophorin C
View Full Product Details