Product Details

SNP ID
rs13480902
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.11:19989571 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GCACTATAAGGGAGGCGCAGCATGA[C/G]TTCCATGCTCCGTGCTTGTCGCACC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
Spred2 PubMed Links

Gene Details

Gene
Spred2
Gene Name
sprouty-related, EVH1 domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033523.4 Intron NP_277058.1

View Full Product Details