Product Details

SNP ID
rs13476984
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.3:14131979 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
ACATGGGGGAAGTTCTGAGGGAGCT[A/G]CATGAGGTTTGTTTCGCCATTGACT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Ralyl PubMed Links

Gene Details

Gene
Ralyl
Gene Name
RALY RNA binding protein-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001163328.1 Intron NP_001156800.1
NM_001163329.1 Intron NP_001156801.1
NM_001163330.1 Intron NP_001156802.1
NM_178631.4 Intron NP_848746.1

View Full Product Details