Product Details

SNP ID
rs13483547
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.19:17477616 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TCTATTATTAAAGCCTGTCCATACA[A/G]AGCAAAATTTGAATGGGGGTGACCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Pcsk5 PubMed Links

Gene Details

Gene
Pcsk5
Gene Name
proprotein convertase subtilisin/kexin type 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001163144.1 Intron NP_001156616.1
NM_001190483.1 Intron NP_001177412.1

View Full Product Details