Product Details

SNP ID
rs6207385
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.15:44577835 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
AAAATGTCTTTTATGTAACCCACAG[A/T]GTATATAATGTAAACAAAGAGGCCA
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
Pkhd1l1 PubMed Links

Gene Details

Gene
Pkhd1l1
Gene Name
polycystic kidney and hepatic disease 1-like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138674.2 Intron NP_619615.2

View Full Product Details