Product Details

SNP ID
rs13481371
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.12:30198600 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GCTAGATTAAATGTTAAGACCTGTC[A/G]TCAGAGCCCCATTTGAGACCTACCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Sntg2 PubMed Links

Gene Details

Gene
Sntg2
Gene Name
syntrophin, gamma 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_172951.3 Intron NP_766539.2

View Full Product Details