Product Details

SNP ID
rs6244850
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.14:69949425 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TACTTCCTTATGAAGAAGACACAAG[A/C]CCCTTTCACCCTGACCCCCGCCCTG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
Pebp4 PubMed Links

Gene Details

Gene
Pebp4
Gene Name
phosphatidylethanolamine binding protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_028526.4 Intron NP_082802.1
NM_028560.4 Intron NP_082836.2

View Full Product Details