Product Details

SNP ID
rs3689848
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.15:34840082 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CTTTTGCAATGTTTAGAGAGGTTTC[T/C]TTTTTCTTTTGAGGATGGTGTGTCT
Phenotype
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
Gm32970 PubMed Links

Gene Details

Gene
Gm32970
Gene Name
predicted gene, 32970
There are no transcripts associated with this gene.

Gene
Kcns2
Gene Name
K+ voltage-gated channel, subfamily S, 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271704.1 1763 UTR 3 NP_001258633.1
NM_181317.4 1763 UTR 3 NP_851834.1

View Full Product Details