Product Details

SNP ID
rs4187972
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.16:55844692 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GGTCCACTGCAAGGAAGGGACCGAC[A/G]TTCTTGGGACTGCCCAAATTGAACT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Nfkbiz PubMed Links

Gene Details

Gene
Nfkbiz
Gene Name
nuclear factor of kappa light polypeptide gene enhancer in B cells inhibitor, zeta
There are no transcripts associated with this gene.

Gene
Nxpe3
Gene Name
neurexophilin and PC-esterase domain family, member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134457.1 1691 Silent Mutation NP_001127929.1

View Full Product Details