Product Details

SNP ID
rs13481939
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.13:90883686 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
AGTGGTCAGGAGAGGTGATAAACTC[A/G]TAATGACGGTCCAGATAGCGCAGGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Atp6ap1l PubMed Links

Gene Details

Gene
Atp6ap1l
Gene Name
ATPase, H+ transporting, lysosomal accessory protein 1-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145879.1 939 Silent Mutation NP_001139351.1

View Full Product Details