Product Details

SNP ID
rs13481127
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.11:83412529 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
AGGCAAGCAGAGGGAGCAATGGTTG[G/T]TCCTGACGGTGGCTGAGCCCCCAGC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
Ap2b1 PubMed Links

Gene Details

Gene
Ap2b1
Gene Name
adaptor-related protein complex 2, beta 1 subunit
There are no transcripts associated with this gene.

Gene
Gas2l2
Gene Name
growth arrest-specific 2 like 2
There are no transcripts associated with this gene.

Gene
Rasl10b
Gene Name
RAS-like, family 10, member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013386.2 572 UTR 5 NP_001013404.1

View Full Product Details