Product Details
- SNP ID
-
rs13475820
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:38517287 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAAAAAGGTCAGTGGGAATGACACT[A/G]TAAAGCAGGAAGGGAGAACCAGGAT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Aff3
PubMed Links
Gene Details
- Gene
- Aff3
- Gene Name
- AF4/FMR2 family, member 3
There are no transcripts associated with this gene.
View Full Product Details