Product Details
- SNP ID
-
rs4230429
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:69697562 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGCCACCAGCCTGCGGGCCACCGC[T/C]GAATCTGTCTGCGGCCTCTCCTTCG
- Phenotype
-
- Polymorphism
- T/C, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Loxl2
PubMed Links
Gene Details
- Gene
- Loxl2
- Gene Name
- lysyl oxidase-like 2
There are no transcripts associated with this gene.
- Gene
- Mir6950
- Gene Name
- microRNA 6950
There are no transcripts associated with this gene.
- Gene
- R3hcc1
- Gene Name
- R3H domain and coiled-coil containing 1
There are no transcripts associated with this gene.
View Full Product Details