Product Details

SNP ID
rs3725809
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.14:64248634 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
ATCAAACTTAACAGTAAACCGAGCC[A/G]AGTGATACCCATGAGAATGCTAAAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Msra PubMed Links

Gene Details

Gene
Msra
Gene Name
methionine sulfoxide reductase A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001253712.1 Intron NP_001240641.1
NM_001253714.1 Intron NP_001240643.1
NM_001253715.1 Intron NP_001240644.1
NM_001253716.1 Intron NP_001240645.1
NM_026322.4 Intron NP_080598.2

View Full Product Details