Product Details
- SNP ID
-
rs13480667
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:91093712 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CATCCAGTCAACGAAGCCTGACACT[A/G]TGACTAGTTACAAGTGTTTCTCAGA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Ikbip
PubMed Links
Gene Details
- Gene
- Ikbip
- Gene Name
- IKBKB interacting protein
View Full Product Details