Product Details

SNP ID
rs13476303
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.1:191069703 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TCCCCGAGGTCCTTCTCCCTGAGCG[A/T]TCTCATCCAGTGCTTTGTCCGCACC
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
Nsl1 PubMed Links

Gene Details

Gene
Nsl1
Gene Name
NSL1, MIND kinetochore complex component, homolog (S. cerevisiae)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198654.3 Intron NP_941056.3
Gene
Tatdn3
Gene Name
TatD DNase domain containing 3
There are no transcripts associated with this gene.

View Full Product Details