Product Details
- SNP ID
-
rs13476290
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:187169478 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAACTATAAGGACAAAAAGATTAAA[A/G]TGTTCTCCAGTCCCTTCCAATAATA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Spata17
PubMed Links
Gene Details
- Gene
- Spata17
- Gene Name
- spermatogenesis associated 17
View Full Product Details