Product Details

SNP ID
rs13481520
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.12:71318147 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CAAGGAGCGCCACCTTGAAGGCCCG[C/T]GGCCAGGCAGCCATGGAACACGGCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
2700049A03Rik PubMed Links

Gene Details

Gene
2700049A03Rik
Gene Name
RIKEN cDNA 2700049A03 gene
There are no transcripts associated with this gene.

Gene
Dact1
Gene Name
dapper homolog 1, antagonist of beta-catenin (xenopus)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190466.1 1801 Silent Mutation NP_001177395.1
NM_021532.4 1801 Silent Mutation NP_067507.2

View Full Product Details