Product Details
- SNP ID
-
rs13477177
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:69429568 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAGCCAGGTCACTGTGTGTGTGCGC[A/G]CGCGCTTTAAGGGGCCCTCAGGACC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Ppm1l
PubMed Links
Gene Details
- Gene
- Ppm1l
- Gene Name
- protein phosphatase 1 (formerly 2C)-like
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_178726.3 |
|
Intron |
|
|
NP_848841.2 |
View Full Product Details