Product Details
- SNP ID
-
rs13476121
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:134670008 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTCCTCTAGGTTCCACCCCTAAAGC[C/T]GCTGCCACCTCTCATGAGCACCCCT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Syt2
PubMed Links
Gene Details
- Gene
- Syt2
- Gene Name
- synaptotagmin II
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_009307.3 |
|
Intron |
|
|
NP_033333.2 |
View Full Product Details