Product Details

SNP ID
rs13459075
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.4:41717393 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GCGCTGTCCAGCACAGGCACCAGGG[A/C]GTTCACTTGTTCCAGGAGTGCCACC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
Arid3c PubMed Links

Gene Details

Gene
Arid3c
Gene Name
AT rich interactive domain 3C (BRIGHT-like)
There are no transcripts associated with this gene.

Gene
Dctn3
Gene Name
dynactin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159565.1 349 Missense Mutation NP_001153037.1
NM_016890.4 349 Missense Mutation NP_058586.3
Gene
Rpp25l
Gene Name
ribonuclease P/MRP 25 subunit-like
There are no transcripts associated with this gene.

View Full Product Details