Product Details
- SNP ID
-
rs13476254
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:175152896 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCCAGTAAGTCACTACCTGAAATTT[C/T]CTTAAGGTCTCTTATGTTGAAATAT
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
Rgs7
PubMed Links
Gene Details
- Gene
- Rgs7
- Gene Name
- regulator of G protein signaling 7
View Full Product Details