Product Details

SNP ID
rs13483244
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.18:20824260 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TTGTAACAACTCTGTGACTACCTTT[A/G]ATTTTAAGTTTGAAAACTGCATTAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Trappc8 PubMed Links

Gene Details

Gene
Trappc8
Gene Name
trafficking protein particle complex 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_029491.2 Intron NP_083767.2
NM_177038.3 Intron NP_796012.2

View Full Product Details