Product Details
- SNP ID
-
rs3721417
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:25633874 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTGATAACAGCAACCTGAAAGTAGT[G/T]TCAAAGAGTCAACATTCTTCTGTGT
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
Fut9
PubMed Links
Gene Details
- Gene
- Fut9
- Gene Name
- fucosyltransferase 9
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_010243.3 |
|
Intron |
|
|
NP_034373.1 |
View Full Product Details