Product Details

SNP ID
rs13480479
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.10:7644364 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
AGGCGGTGAGGATTCCACTTCCAGA[G/T]CCCACGTCCAGGGCTTTAGCACCTT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
Pcmt1 PubMed Links

Gene Details

Gene
Pcmt1
Gene Name
protein-L-isoaspartate (D-aspartate) O-methyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_008786.2 713 Silent Mutation NP_032812.2

View Full Product Details