Product Details
- SNP ID
-
rs6392741
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:17900220 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAAACCTATTGTGCCCATTTTGCTT[C/T]TGTCTTATTCATGTATTTTGTCTAC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Mmp16
PubMed Links
Gene Details
- Gene
- Mmp16
- Gene Name
- matrix metallopeptidase 16
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_019724.3 |
|
Intron |
|
|
NP_062698.2 |
View Full Product Details