Product Details
- SNP ID
-
rs13476894
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:166051427 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAGACGTGAATGCCAGTCCCGAAAC[A/G]CGCCAGAGATATGAAACAATGCAGT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Ncoa3
PubMed Links
Gene Details
- Gene
- Ncoa3
- Gene Name
- nuclear receptor coactivator 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_008679.3 |
871 |
Silent Mutation |
|
|
NP_032705.2 |
View Full Product Details