Product Details

SNP ID
rs13475815
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.1:37124649 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CAGTATCTTTCCTTTGAGGGCTCGA[C/T]CTAAACATGCCTCTGGGCAACATCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Gm37020 PubMed Links

Gene Details

Gene
Gm37020
Gene Name
predicted gene, 37020
There are no transcripts associated with this gene.

Gene
Vwa3b
Gene Name
von Willebrand factor A domain containing 3B
There are no transcripts associated with this gene.

View Full Product Details