Product Details
- SNP ID
-
rs13478550
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:137587089 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCTCAGTGGCGACGTTTGGAACAT[C/T]GACAATAACTTTTGAAGCCAAAAGC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Mospd3
PubMed Links
Gene Details
- Gene
- Mospd3
- Gene Name
- motile sperm domain containing 3
There are no transcripts associated with this gene.
- Gene
- Tfr2
- Gene Name
- transferrin receptor 2
There are no transcripts associated with this gene.
View Full Product Details