Product Details

SNP ID
rs13478128
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.5:20323281 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CCAAATCCATGCTGGGCTTAGCCAA[A/C]GCTTCAGCTGTGCCTCCCAACTTCA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
Magi2 PubMed Links

Gene Details

Gene
Magi2
Gene Name
membrane associated guanylate kinase, WW and PDZ domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170745.1 Intron NP_001164216.1
NM_001170746.1 Intron NP_001164217.1
NM_015823.3 Intron NP_056638.1

View Full Product Details