Product Details

SNP ID
rs13478697
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.6:32850650 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GGAATATTTTCCTTCTGGACATCTT[C/T]CTCTTGCACCTACTCAATGCCTTTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Chchd3 PubMed Links

Gene Details

Gene
Chchd3
Gene Name
coiled-coil-helix-coiled-coil-helix domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025336.1 Intron NP_079612.1

View Full Product Details